Learn More
Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate | 653-63-4 | 99.2% | 331.22 g/mol | C10H14N5O6P | 50 MG
SDP
Catalog No. 5000303131
Encompass_Preferred
2'-Deoxyadenosine-5'-monophosphate is a deoxyadenosine monophosphate reference standard used in biochemical and analytical workflows. Supplied as a white to off-white solid with high purity, it is intended for method development, calibration, and quality control in nucleic acid research and mass spectrometry applications.
  • High purity (99.2%) for reliable analytical results.
  • Molecular weight 331.22 g/mol and formula C10H14N5O6P for accurate mass calculations.
  • Suitable as a calibration standard for LC-MS and HPLC methods.
  • Stable solid form for convenient storage and handling.
  • Available in small laboratory-scale quantities for research use.
Catalog No. 50-003-03131 Supplier Medchemexpress LLC Supplier No. HYW016009R50MG
2'-Deoxyadenosine-5'-monophosphate is a deoxyadenosine monophosphate reference standard used in biochemical and analytical workflows. Supplied as a white to off-white solid with high purity, it is intended for method development, calibration, and quality control in nucleic acid research and mass spectrometry applications.
  • High purity (99.2%) for reliable analytical results.
  • Molecular weight 331.22 g/mol and formula C10H14N5O6P for accurate mass calculations.
  • Suitable as a calibration standard for LC-MS and HPLC methods.
  • Stable solid form for convenient storage and handling.
  • Available in small laboratory-scale quantities for research use.