Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate | 653-63-4 | MFCD00005753 | 99.6% | 331.22 g/mol | C10H14N5O6P | 1 ML
SDP

Catalog No. 5000300279
Encompass_Preferred

2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxynucleotide used in biochemical and molecular biology research as an adenine-containing monophosphate component of DNA. It is used to study DNA synthesis, nucleotide interactions, and DNA damage/repair. The compound is supplied as a solid and as a ready-to-use 10 mM solution in DMSO for assay use.

  • Adenine nucleoside monophosphate for DNA research.
  • Available as solid or 10 mM solution in DMSO for immediate use.
  • High purity suitable for biochemical assays.
  • Compatible with DNA synthesis and damage/repair studies.
  • Formula C10H14N5O6P and molecular weight 331.22 g/mol; CAS 653-63-4 for identification.

Catalog No. 50-003-00279 Supplier Medchemexpress LLC Supplier No. HYW01600910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxynucleotide used in biochemical and molecular biology research as an adenine-containing monophosphate component of DNA. It is used to study DNA synthesis, nucleotide interactions, and DNA damage/repair. The compound is supplied as a solid and as a ready-to-use 10 mM solution in DMSO for assay use.

  • Adenine nucleoside monophosphate for DNA research.
  • Available as solid or 10 mM solution in DMSO for immediate use.
  • High purity suitable for biochemical assays.
  • Compatible with DNA synthesis and damage/repair studies.
  • Formula C10H14N5O6P and molecular weight 331.22 g/mol; CAS 653-63-4 for identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.