Learn More
Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate-d12 dilithium | 97.7% | 355.16 | C10D12Li2N5O6P | 1 MG
SDP
Catalog No. 5000300280
Encompass_Preferred
2'-Deoxyadenosine-5'-monophosphate-d12 (dilithium) is a deuterium-labeled form of 2'-deoxyadenosine-5'-monophosphate used as a research reagent to study DNA synthesis, nucleotide interactions, and isotope tracing by mass spectrometry or NMR. Supplied as the dilithium salt, it is intended for laboratory research use only.
  • Isotopically labeled d12 for accurate mass-shift studies.
  • Suitable for mass spectrometry and NMR applications.
  • Dilithium salt form for improved solubility in selected buffers.
  • High purity (97.7%) for reproducible experimental results.
  • Compatible with nucleotide chemistry and DNA interaction studies.
Catalog No. 50-003-00280 Supplier Medchemexpress LLC Supplier No. HYW016009S1MG
2'-Deoxyadenosine-5'-monophosphate-d12 (dilithium) is a deuterium-labeled form of 2'-deoxyadenosine-5'-monophosphate used as a research reagent to study DNA synthesis, nucleotide interactions, and isotope tracing by mass spectrometry or NMR. Supplied as the dilithium salt, it is intended for laboratory research use only.
  • Isotopically labeled d12 for accurate mass-shift studies.
  • Suitable for mass spectrometry and NMR applications.
  • Dilithium salt form for improved solubility in selected buffers.
  • High purity (97.7%) for reproducible experimental results.
  • Compatible with nucleotide chemistry and DNA interaction studies.