Learn More
Abcam 2'-Deo x yadenosine 5'-triphosphate [dATP] trisodium salt trihydrate, nucleoside triphosphate, 50MG
Catalog No. 502704582
Encompass_Preferred
MW 611.13 g/mol, Purity >98%. Nucleoside triphosphate. Substrate for a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s). Used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4582 Supplier Abcam Supplier No. AB14653750MG
MW 611.13 g/mol, Purity >98%. Nucleoside triphosphate. Substrate for a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s). Used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
The product is subject to the following: Abcam Restricted Use Statement