Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate-13C10,15N5 (dilithium) | 1927-31-7 | 99.2% | 517.94 | 1 MG
SDP
Catalog No. 5000348333
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate-13C10,15N5 (dATP-13C10,15N5) dilithium is a 13C and 15N-labeled nucleotide essential for DNA synthesis as a substrate of DNA polymerase. It is valuable in research as:
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48333 Supplier Medchemexpress LLC Supplier No. HY136648S11MG
2'-Deoxyadenosine-5'-triphosphate-13C10,15N5 (dATP-13C10,15N5) dilithium is a 13C and 15N-labeled nucleotide essential for DNA synthesis as a substrate of DNA polymerase. It is valuable in research as:
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.