Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate-13C10 dilithium | 1927-31-7 | 95.0% | 512.97 | 1 MG
SDP
Catalog No. 5000349210
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate-13C10 dilithium (dATP-13C10 dilithium) is a 13C-labeled version of 2'-Deoxyadenosine-5'-triphosphate (dATP). dATP is a nucleotide crucial for DNA synthesis or replication in cells, serving as a substrate for DNA polymerase.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for analysis by NMR
  • Suitable for analysis by GC-MS
  • Suitable for analysis by LC-MS
Catalog No. 50-003-49210 Supplier Medchemexpress LLC Supplier No. HY136648S51MG
2'-Deoxyadenosine-5'-triphosphate-13C10 dilithium (dATP-13C10 dilithium) is a 13C-labeled version of 2'-Deoxyadenosine-5'-triphosphate (dATP). dATP is a nucleotide crucial for DNA synthesis or replication in cells, serving as a substrate for DNA polymerase.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for analysis by NMR
  • Suitable for analysis by GC-MS
  • Suitable for analysis by LC-MS