Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate-15N5, dilithium | 99.3% | 508.01 | 1 MG
SDP

Catalog No. 5000348658
Encompass_Preferred

This product is 2'-Deoxyadenosine-5'-triphosphate-15N5, dilithium, a chemical compound with a molecular weight of 508.01. It is supplied as a colorless to light yellow liquid solution and has a purity of 99.3%. This compound is intended for research use only.

  • High purity of 99.3%
  • Isotopic enrichment of 98.7%
  • Available as a colorless to light yellow liquid
  • Store at -20°C for up to 2 years
  • Molecular formula C10H14Li215N5O12P3

Catalog No. 50-003-48658 Supplier Medchemexpress LLC Supplier No. HY136648S41MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is 2'-Deoxyadenosine-5'-triphosphate-15N5, dilithium, a chemical compound with a molecular weight of 508.01. It is supplied as a colorless to light yellow liquid solution and has a purity of 99.3%. This compound is intended for research use only.

  • High purity of 99.3%
  • Isotopic enrichment of 98.7%
  • Available as a colorless to light yellow liquid
  • Store at -20°C for up to 2 years
  • Molecular formula C10H14Li215N5O12P3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.