Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate-d14 (dilithium) (dATP-d14 (dilithium)) | 2687960-68-3 | 97.10% | 517.13 | 1 MG
SDP
Catalog No. 5000348757
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate-d14 (dATP-d14) dilithium is deuterium labeled 2'-Deoxyadenosine-5'-triphosphate. It is a nucleotide used in cells for DNA synthesis (or replication), as a substrate of DNA polymerase. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48757 Supplier Medchemexpress LLC Supplier No. HY136648S31MG
2'-Deoxyadenosine-5'-triphosphate-d14 (dATP-d14) dilithium is deuterium labeled 2'-Deoxyadenosine-5'-triphosphate. It is a nucleotide used in cells for DNA synthesis (or replication), as a substrate of DNA polymerase. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.