Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxyadenosine-5'-triphosphate disodium salt, 98%, Thermo Scientific™

Catalog No. AAJ64045MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64045MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64045MC Supplier Thermo Scientific Chemicals Supplier No. J64045MC

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Deoxyadenosine-5′-triphosphate disodium salt is used as a substrate by a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s).

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74299-50-6
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.15
MDL Number MFCD00080335
InChI Key JEKDCIBJADJZSK-DZVPOJRENA-L
Synonym 2'-deoxyadenosine-5'-triphosphate disodium salt
PubChem CID 131674273
IUPAC Name disodium;[[[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 535.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-triphosphate disodium salt
EINECSNumber 277-809-2
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.