Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate (trisodium) | 54680-12-5 | 98.0% | 557.13 | 50 MG
SDP
Catalog No. 5000350719
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate (dATP) trisodium is a nucleotide used in cells for DNA synthesis (or replication), as a substrate of DNA polymerase. It is promising for research of genetic immunodeficiency diseases adenosine deaminase deficiency and purine nucleotide phosphorylase deficiency.
  • Used as a nucleotide for DNA synthesis (replication).
  • Substrate of DNA polymerase.
  • Promising for research in genetic immunodeficiency diseases (adenosine deaminase deficiency and purine nucleotide phosphorylase deficiency).
  • Induces cytochrome C release in mouse liver mitochondria (0.5 mM; 0-10 min).
  • Stimulates bacterial biofilm formation in vitro and bacterial adherence to human epithelial cells (400 μM, 16 h).
Catalog No. 50-003-50719 Supplier Medchemexpress LLC Supplier No. HY136648A50MG
Add to Cart
Add to Cart
2'-Deoxyadenosine-5'-triphosphate (dATP) trisodium is a nucleotide used in cells for DNA synthesis (or replication), as a substrate of DNA polymerase. It is promising for research of genetic immunodeficiency diseases adenosine deaminase deficiency and purine nucleotide phosphorylase deficiency.
  • Used as a nucleotide for DNA synthesis (replication).
  • Substrate of DNA polymerase.
  • Promising for research in genetic immunodeficiency diseases (adenosine deaminase deficiency and purine nucleotide phosphorylase deficiency).
  • Induces cytochrome C release in mouse liver mitochondria (0.5 mM; 0-10 min).
  • Stimulates bacterial biofilm formation in vitro and bacterial adherence to human epithelial cells (400 μM, 16 h).