Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate trisodium | 54680-12-5 | 99.0% | 557.13 | 5 ML
SDP
Catalog No. 5000351888
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate trisodium is a raw material used by DNA polymerase to synthesize long DNA chains during DNA replication. It is also utilized in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is provided as an aqueous solution.
  • Raw material for DNA synthesis
  • Utilized in reverse transcription polymerase chain reaction (RT-PCR)
  • Provided as an aqueous solution
Catalog No. 50-003-51888 Supplier Medchemexpress LLC Supplier No. HY136648B5ML
2'-Deoxyadenosine-5'-triphosphate trisodium is a raw material used by DNA polymerase to synthesize long DNA chains during DNA replication. It is also utilized in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is provided as an aqueous solution.
  • Raw material for DNA synthesis
  • Utilized in reverse transcription polymerase chain reaction (RT-PCR)
  • Provided as an aqueous solution