Learn More
Medchemexpress LLC 2'-Deoxyadenosine-5'-triphosphate (trisodium), PCR grade | 54680-12-5 | PCR Grade | 557.13 | 1 ML
SDP
Catalog No. 5000350721
Encompass_Preferred
2'-Deoxyadenosine-5'-triphosphate trisodium, 100 mM solution, PCR Grade, is a nucleotide utilized by cells for DNA synthesis and replication. It serves as a raw material for DNA polymerase to synthesize long DNA strands and is also used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is supplied as an aqueous solution.
  • Substrate for DNA polymerase
  • Facilitates DNA synthesis and replication
  • Suitable for reverse transcription polymerase chain reaction (RT-PCR)
  • Aqueous solution format
Catalog No. 50-003-50721 Supplier Medchemexpress LLC Supplier No. HY136648B1ML
2'-Deoxyadenosine-5'-triphosphate trisodium, 100 mM solution, PCR Grade, is a nucleotide utilized by cells for DNA synthesis and replication. It serves as a raw material for DNA polymerase to synthesize long DNA strands and is also used in reverse transcription polymerase chain reaction (RT-PCR) for DNA amplification. This product is supplied as an aqueous solution.
  • Substrate for DNA polymerase
  • Facilitates DNA synthesis and replication
  • Suitable for reverse transcription polymerase chain reaction (RT-PCR)
  • Aqueous solution format