Learn More
Medchemexpress LLC 2'-deoxyadenosine | 958-09-8 | MFCD00005754 | 251.24 g/mol | C10H13N5O3 | 50 MG
SDP
Catalog No. 5000209114
Encompass_Preferred
2'-Deoxyadenosine is an adenine nucleoside used as a research reagent and analytical standard. It inhibits glucose-stimulated insulin release, modulates islet cAMP, activates caspase-3, and has been reported to inhibit S-adenosyl-L-homocysteine hydrolase; it is used in analytical and biochemical studies.
  • intended for research and analytical applications
  • inhibits glucose-stimulated insulin release
  • modulates islet cyclic AMP and activates caspase-3
  • reported inhibitor of S-adenosyl-L-homocysteine hydrolase
  • available in labeled and standard variants for analytical use
Catalog No. 50-002-09114 Supplier Medchemexpress LLC Supplier No. HYW040329R50MG
Add to Cart
Add to Cart
2'-Deoxyadenosine is an adenine nucleoside used as a research reagent and analytical standard. It inhibits glucose-stimulated insulin release, modulates islet cAMP, activates caspase-3, and has been reported to inhibit S-adenosyl-L-homocysteine hydrolase; it is used in analytical and biochemical studies.
  • intended for research and analytical applications
  • inhibits glucose-stimulated insulin release
  • modulates islet cyclic AMP and activates caspase-3
  • reported inhibitor of S-adenosyl-L-homocysteine hydrolase
  • available in labeled and standard variants for analytical use