Learn More
Medchemexpress LLC 2'-deoxyadenosine | 958-09-8 | MFCD00005754 | 251.24 g/mol | C10H13N5O3 | 500 MG
SDP
Catalog No. 5000209113
Encompass_Preferred
2'-deoxyadenosine is an adenine nucleoside supplied for research and analytical applications. The compound has been reported to inhibit glucose-stimulated insulin release, activate caspase-3 and promote apoptosis, and inhibit S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Adenine nucleoside for research applications.
  • Modulates insulin-stimulated cAMP accumulation and insulin release.
  • Promotes apoptosis via caspase-3 activation.
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Available from suppliers in small research quantities (supplier pack indicated as 500 MG in provided row data).
Catalog No. 50-002-09113 Supplier Medchemexpress LLC Supplier No. HYW040329500MG
2'-deoxyadenosine is an adenine nucleoside supplied for research and analytical applications. The compound has been reported to inhibit glucose-stimulated insulin release, activate caspase-3 and promote apoptosis, and inhibit S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Adenine nucleoside for research applications.
  • Modulates insulin-stimulated cAMP accumulation and insulin release.
  • Promotes apoptosis via caspase-3 activation.
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Available from suppliers in small research quantities (supplier pack indicated as 500 MG in provided row data).