Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyadenosine | 958-09-8 | MFCD00005754 | 99.9% | 251.24 | C10H13N5O3 | 25 MG
SDP

Catalog No. 5000209075
Encompass_Preferred

2'-deoxyadenosine is an analytical standard of the adenine nucleoside 2'-deoxyadenosine supplied for research and analytical use. It inhibits glucose-stimulated insulin release and cAMP accumulation, activates caspase-3 to promote apoptosis, and shows antiproliferative effects in some cell models. It is intended for use as a reference in HPLC, GC, and MS methods and is not for human use.

  • Analytical standard for HPLC, GC, and MS applications.
  • High purity, 99.9% (as supplied in COA).
  • CAS number 958-09-8 for unambiguous identification.
  • Molecular formula C10H13N5O3; molecular weight 251.24.
  • Available in small pack sizes suitable for method development and reference standards.

Catalog No. 50-002-09075 Supplier Medchemexpress LLC Supplier No. 25MGHYW040329R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-deoxyadenosine is an analytical standard of the adenine nucleoside 2'-deoxyadenosine supplied for research and analytical use. It inhibits glucose-stimulated insulin release and cAMP accumulation, activates caspase-3 to promote apoptosis, and shows antiproliferative effects in some cell models. It is intended for use as a reference in HPLC, GC, and MS methods and is not for human use.

  • Analytical standard for HPLC, GC, and MS applications.
  • High purity, 99.9% (as supplied in COA).
  • CAS number 958-09-8 for unambiguous identification.
  • Molecular formula C10H13N5O3; molecular weight 251.24.
  • Available in small pack sizes suitable for method development and reference standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.