Learn More
Medchemexpress LLC 2'-deoxyadenosine | 958-09-8 | MFCD00005754 | 99.9% | 251.24 g/mol | C10H13N5O3 | 1 ML
SDP
Catalog No. 5000208508
Encompass_Preferred
2'-deoxyadenosine solution (10 mM in DMSO, 1 mL) is an adenine nucleoside supplied as a ready-to-use research solution intended for biochemical and analytical applications. It is commonly used as an analytical standard and in studies of nucleoside metabolism, apoptosis, and cell growth inhibition.
  • Ready-to-use 10 mM solution in DMSO.
  • Net volume: 1 mL.
  • Intended for research and analytical applications.
  • Used in studies of nucleoside metabolism, insulin secretion, and apoptosis.
  • High purity reported for the solid standard (99.94%).
Catalog No. 50-002-08508 Supplier Medchemexpress LLC Supplier No. HYW04032910MM/1ML
2'-deoxyadenosine solution (10 mM in DMSO, 1 mL) is an adenine nucleoside supplied as a ready-to-use research solution intended for biochemical and analytical applications. It is commonly used as an analytical standard and in studies of nucleoside metabolism, apoptosis, and cell growth inhibition.
  • Ready-to-use 10 mM solution in DMSO.
  • Net volume: 1 mL.
  • Intended for research and analytical applications.
  • Used in studies of nucleoside metabolism, insulin secretion, and apoptosis.
  • High purity reported for the solid standard (99.94%).