Learn More
Apexbio Technology LLC 2-Deoxyadenosine-monoH2O
SDP
Catalog No. 5000569774
Encompass_Preferred
2 -Deoxyadenosine monohydrate (CAS No 16373-93-6) is widely used in biomedical research particularly in molecular biology and pharmacology studies This compound originates from the deoxyribonucleic acid (DNA) and acts by integrating into DNA strands impacting replication and cellular processes It targets pathways involved in nucleoside metabolism and DNA synthesis In vitro it demonstrates activity typically in the nanomolar to low micromolar range making it valuable for studying DNA replication and repair mechanisms Common applications include cell-based assays animal studies and drug screening to investigate its effects on cellular proliferation and apoptosis Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69774 Supplier Apexbio Technology LLC Supplier No. C7028100
2 -Deoxyadenosine monohydrate (CAS No 16373-93-6) is widely used in biomedical research particularly in molecular biology and pharmacology studies This compound originates from the deoxyribonucleic acid (DNA) and acts by integrating into DNA strands impacting replication and cellular processes It targets pathways involved in nucleoside metabolism and DNA synthesis In vitro it demonstrates activity typically in the nanomolar to low micromolar range making it valuable for studying DNA replication and repair mechanisms Common applications include cell-based assays animal studies and drug screening to investigate its effects on cellular proliferation and apoptosis Experimental concentrations typically depend on the specific research design