Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyadenosine monohydrate-13C10,15N5 hydrate | 2483830-27-7 | 99.7% | 284.15 | 1 MG
SDP

Catalog No. 5000452574
Encompass_Preferred

2'-Deoxyadenosine monohydrate-13C10,15N5 hydrate | 2483830-27-7 | 99.7% | 284.15 | 1 MG

Catalog No. 50-004-52574 Supplier Medchemexpress LLC Supplier No. HYW011683S61MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyadenosine monohydrate-13C10,15N5 hydrate | 2483830-27-7 | 99.7% | 284.15 | 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.