Learn More
Medchemexpress LLC 2'-Deoxyadenosine monohydrate | 16373-93-6 | 99.89% | 269.26 | 5 G
SDP
Catalog No. 5000452325
Encompass_Preferred
2'-Deoxyadenosine monohydrate is an adenine nucleoside with multiple biological roles, including inhibiting glucose-stimulated insulin release and islet cyclic AMP (cAMP) accumulation. It activates caspase-3 and promotes apoptosis, and inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity. It has shown an anticancer effect on colon cancer and appears as a white to off-white solid.
  • Inhibits glucose-stimulated insulin release
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation
  • Activates caspase-3
  • Promotes apoptosis
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity
  • Demonstrated inhibitory effect on the growth of various cells, including anticancer effect on colon cancer
  • Appears as a white to off-white solid
Catalog No. 50-004-52325 Supplier Medchemexpress LLC Supplier No. HYW0116835G
2'-Deoxyadenosine monohydrate is an adenine nucleoside with multiple biological roles, including inhibiting glucose-stimulated insulin release and islet cyclic AMP (cAMP) accumulation. It activates caspase-3 and promotes apoptosis, and inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity. It has shown an anticancer effect on colon cancer and appears as a white to off-white solid.
  • Inhibits glucose-stimulated insulin release
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation
  • Activates caspase-3
  • Promotes apoptosis
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity
  • Demonstrated inhibitory effect on the growth of various cells, including anticancer effect on colon cancer
  • Appears as a white to off-white solid