Learn More
Medchemexpress LLC 2'-Deoxyadenosine monohydrate | 16373-93-6 | 99.9% | 269.26 | 1 ML
SDP
Catalog No. 5000452426
Encompass_Preferred
2'-Deoxyadenosine monohydrate is an adenine nucleoside used in research that demonstrates various biological activities including the inhibition of glucose-stimulated insulin release and islet cAMP accumulation. It also activates caspase-3, promotes apoptosis, inhibits SAHH activity, and has an anticancer effect on colon cancer by inhibiting cell growth.
  • Inhibits glucose-stimulated insulin release.
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation.
  • Activates caspase-3 and promotes apoptosis.
  • Inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Inhibits the growth of various cells.
  • Has an anticancer effect on colon cancer.
Catalog No. 50-004-52426 Supplier Medchemexpress LLC Supplier No. HYW01168310MM/1ML
2'-Deoxyadenosine monohydrate is an adenine nucleoside used in research that demonstrates various biological activities including the inhibition of glucose-stimulated insulin release and islet cAMP accumulation. It also activates caspase-3, promotes apoptosis, inhibits SAHH activity, and has an anticancer effect on colon cancer by inhibiting cell growth.
  • Inhibits glucose-stimulated insulin release.
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation.
  • Activates caspase-3 and promotes apoptosis.
  • Inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Inhibits the growth of various cells.
  • Has an anticancer effect on colon cancer.