Learn More
Medchemexpress LLC 2'-Deoxyadenosine monohydrate | 16373-93-6 | 99.9% | 269.26 | 10 G
SDP
Catalog No. 5000452324
Encompass_Preferred
2′-Deoxyadenosine monohydrate is an adenine nucleoside that inhibits glucose-stimulated insulin release and glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation. It activates caspase-3 and promotes apoptosis, and inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH). This compound can inhibit the growth of various cells and has an anticancer effect on colon cancer.
  • Inhibits glucose-stimulated insulin release
  • Activates caspase-3 and promotes apoptosis
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity
  • Inhibits the growth of various cells
  • Demonstrates an anticancer effect on colon cancer
  • For research use only
Catalog No. 50-004-52324 Supplier Medchemexpress LLC Supplier No. HYW01168310G
2′-Deoxyadenosine monohydrate is an adenine nucleoside that inhibits glucose-stimulated insulin release and glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation. It activates caspase-3 and promotes apoptosis, and inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH). This compound can inhibit the growth of various cells and has an anticancer effect on colon cancer.
  • Inhibits glucose-stimulated insulin release
  • Activates caspase-3 and promotes apoptosis
  • Inhibits S-adenosyl-L-homocysteine hydrolase (SAHH) activity
  • Inhibits the growth of various cells
  • Demonstrates an anticancer effect on colon cancer
  • For research use only