Learn More
Medchemexpress LLC 2'-Deoxyadenosine monohydrate | 16373-93-6 | 99.9% | 269.26 | 50 G
SDP
Catalog No. 5000452573
Encompass_Preferred
2'-Deoxyadenosine monohydrate is an adenine nucleoside that inhibits glucose-stimulated insulin release. It also inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation, activates caspase-3, promotes apoptosis, and inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH). Additionally, it inhibits the growth of various cells and exhibits an anticancer effect on colon cancer.
  • Inhibits glucose-stimulated insulin release.
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation.
  • Activates caspase-3 and promotes apoptosis.
  • Inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Inhibits the growth of various cells.
  • Has an anticancer effect on colon cancer.
Catalog No. 50-004-52573 Supplier Medchemexpress LLC Supplier No. HYW01168350G
2'-Deoxyadenosine monohydrate is an adenine nucleoside that inhibits glucose-stimulated insulin release. It also inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation, activates caspase-3, promotes apoptosis, and inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH). Additionally, it inhibits the growth of various cells and exhibits an anticancer effect on colon cancer.
  • Inhibits glucose-stimulated insulin release.
  • Inhibits glucose-stimulated increases in islet cyclic AMP (cAMP) accumulation.
  • Activates caspase-3 and promotes apoptosis.
  • Inhibits the activity of S-adenosyl-L-homocysteine hydrolase (SAHH).
  • Inhibits the growth of various cells.
  • Has an anticancer effect on colon cancer.