Learn More
Bioworld 2'-Deoxyadenosine Monohydrate, 10 g
Catalog No. 50255893
Encompass_Preferred
A deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. It is used in comparison studies of the functions of adenosine analogues on various biological processes. Specifications:Ultra Pure GradeEM (259nm, phosphate buffer, pH 7.0): >14,500Appearance: White crystalline powderChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 187-189 °CRTECS Number: AU7358600
Catalog No. 50-255-893 Supplier Bioworld Supplier No. 404900002
A deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. It is used in comparison studies of the functions of adenosine analogues on various biological processes. Specifications:Ultra Pure GradeEM (259nm, phosphate buffer, pH 7.0): >14,500Appearance: White crystalline powderChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 187-189 °CRTECS Number: AU7358600