Learn More
Cayman Chemical 2′-Deoxyadenosine (hydrate), Purity: ≥98%, CAS: 16373-93-6, 5 g
SDP
Catalog No. 502940261
Encompass_Preferred
A deoxyribonucleoside; intermediate in the purine nucleotide degradation pathway; has been used in the characterization of DNA conformations and the synthesis of nucleoside analogs as antiviral agents
Catalog No. 50-294-0261 Supplier Cayman Chemical Supplier No. 273155 G
A deoxyribonucleoside; intermediate in the purine nucleotide degradation pathway; has been used in the characterization of DNA conformations and the synthesis of nucleoside analogs as antiviral agents