Learn More
Cayman Chemical 2'-Deoxycytidine, Purity: ≥98%, CAS: 951-77-9, 1 g
SDP
Catalog No. 502914048
Encompass_Preferred
A deoxyribonucleoside; phosphorylated by deoxycytidine kinase in the cytosol or TK2 in mitochondria to form dCMP; oral administration in combination with deoxythymidine delays disease onset and increases lifespan in a mouse model of TK2 deficiency
This item is not returnable. View return policy
Catalog No. 50-291-4048 Supplier Cayman Chemical Supplier No. 347081 G
A deoxyribonucleoside; phosphorylated by deoxycytidine kinase in the cytosol or TK2 in mitochondria to form dCMP; oral administration in combination with deoxythymidine delays disease onset and increases lifespan in a mouse model of TK2 deficiency