Learn More
Medchemexpress LLC 2'-Deoxycytidine-15N3 | 951-77-9 | 99.7% | 1 MG
SDP
Catalog No. 5000410945
Encompass_Preferred
2'-Deoxycytidine-15N3 is the 15N labeled 2'-Deoxycytidine. 2'-Deoxycytidine, a deoxyribonucleoside, can inhibit biological effects of Bromodeoxyuridine (Brdu). It is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
Catalog No. 50-004-10945 Supplier Medchemexpress LLC Supplier No. HYD0184S21MG
2'-Deoxycytidine-15N3 is the 15N labeled 2'-Deoxycytidine. 2'-Deoxycytidine, a deoxyribonucleoside, can inhibit biological effects of Bromodeoxyuridine (Brdu). It is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.