Learn More
Medchemexpress LLC 2'-Deoxycytidine-15N3 | 951-77-9 | 99.7% | 5 MG
SDP
Catalog No. 5000409923
Encompass_Preferred
2'-Deoxycytidine-15N3 is the 15N labeled form of 2'-Deoxycytidine. 2'-Deoxycytidine, a deoxyribonucleoside, has been shown to inhibit the biological effects of Bromodeoxyuridine (Brdu). Stable heavy isotopes, such as those of hydrogen, carbon, and other elements, are incorporated into drug molecules primarily to serve as tracers for quantitation during the drug development process. Deuteration, in particular, has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.70%.
Catalog No. 50-004-09923 Supplier Medchemexpress LLC Supplier No. HYD0184S25MG
2'-Deoxycytidine-15N3 is the 15N labeled form of 2'-Deoxycytidine. 2'-Deoxycytidine, a deoxyribonucleoside, has been shown to inhibit the biological effects of Bromodeoxyuridine (Brdu). Stable heavy isotopes, such as those of hydrogen, carbon, and other elements, are incorporated into drug molecules primarily to serve as tracers for quantitation during the drug development process. Deuteration, in particular, has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.70%.