Learn More
Apexbio Technology LLC 2-Deoxycytidine,1g CAS# 951-77-9
SDP
Supplier:  Apexbio Technology LLC B78621000
Encompass_Preferred
2-Deoxycytidine is a cytidine analog [1]. 2-Deoxycytidine prevents DNA methylation by incorporating itself into newly synthesizing DNA strand. 2-Deoxycytidine also binds to DNA methyltransferase irreversibly and hinders its activity. Thus, 2-deoxycytidine was approved as the most efective demethylating agent for the treatment of cancer [1]. 2-Deoxycytidine at clinically achievable and nontoxic concentrations (≥ 100 μmol/L) protected normal bone marrow progenitor cells against the inhibitory effects of co-administered, high concentrations of 3’-azido-3’-deoxythymidine (AZT) (≥ 10 μmol/L). In normal bone marrow mononuclear cells (BMMC), 2-deoxycytidine also significantly corrected AZT-mediated depletion of intracellular thymidine triphosphate and 2-deoxycytidine triphosphate levels. Furthermore, 2-deoxycytidine reduced the intracellular accumulation of AZT triphosphate and its DNA incorporation in BMMC [2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0324