Learn More
TARGETMOL CHEMICALS INC Research Chemical 25mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T497225MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2'-Deoxycytidine-5'-diphosphate trisodium (dCDP) is used as a substrate of CDP (nucleoside diphosphate) kinase (2.7.4.6) for the production of dCTP to support DNA biosynthesis and reverse transcription. purity: 99%
Catalog No. 50-303-8144