Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate | 1032-65-1 | 100.0% | 307.20 | 250 MG
SDP
Catalog No. 5000366410
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis
  • For research use only
Catalog No. 50-003-66410 Supplier Medchemexpress LLC Supplier No. 250MGHYW009216
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis
  • For research use only