Learn More
Apexbio Technology LLC 2-Deoxycytidine 5-monophosphate 1032-65-1 25mg
SDP
Catalog No. 5000091695
Encompass_Preferred
2-Deoxycytidine 5-monophosphate (CAS 1032-65-1) is a deoxycytosine nucleotide with a phosphate group linked to the 5 position of the deoxyribose sugar It serves as a fundamental building block in DNA synthesis participating in enzymatic processes involving DNA polymerases and repair enzymes This compound is widely utilized in studies of nucleic acid metabolism DNA replication and repair mechanisms Its incorporation into oligonucleotides and use in in vitro assays make it valuable for elucidating DNA-protein interactions and the dynamics of nucleic acid biochemistry
Catalog No. 50-000-91695 Supplier Apexbio Technology LLC Supplier No. B786325
2-Deoxycytidine 5-monophosphate (CAS 1032-65-1) is a deoxycytosine nucleotide with a phosphate group linked to the 5 position of the deoxyribose sugar It serves as a fundamental building block in DNA synthesis participating in enzymatic processes involving DNA polymerases and repair enzymes This compound is widely utilized in studies of nucleic acid metabolism DNA replication and repair mechanisms Its incorporation into oligonucleotides and use in in vitro assays make it valuable for elucidating DNA-protein interactions and the dynamics of nucleic acid biochemistry