Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate | 1032-65-1 | 99.6% | 307.20 | 250 MG
SDP

Catalog No. 5000451372
Encompass_Preferred

The product, 2'-Deoxycytidine-5'-monophosphate (2'-Deoxycytidine-5'-monophosphoric acid), is an endogenous metabolite and a deoxynucleotide building block of DNA. It can serve as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thereby supporting DNA biosynthesis.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light, stored under nitrogen
  • In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen)

Catalog No. 50-004-51372 Supplier Medchemexpress LLC Supplier No. 250MGHYW009216R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The product, 2'-Deoxycytidine-5'-monophosphate (2'-Deoxycytidine-5'-monophosphoric acid), is an endogenous metabolite and a deoxynucleotide building block of DNA. It can serve as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thereby supporting DNA biosynthesis.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light, stored under nitrogen
  • In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.