Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate | 1032-65-1 | 99.99% | 307.20 | 50 MG
SDP
Catalog No. 5000452627
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphate (5'-dCMP) is an endogenous metabolite and a deoxynucleotide building block of DNA. It serves as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, which supports DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis
  • Substrate for UMP/CMP kinase
Catalog No. 50-004-52627 Supplier Medchemexpress LLC Supplier No. HYW00921650MG
2'-Deoxycytidine-5'-monophosphate (5'-dCMP) is an endogenous metabolite and a deoxynucleotide building block of DNA. It serves as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, which supports DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis
  • Substrate for UMP/CMP kinase