Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate | 1032-65-1 | MFCD00006546 | ≥95.0% | 1 G
SDP
Catalog No. 5000451665
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be utilized as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thereby supporting DNA biosynthesis.
  • Endogenous metabolite.
  • Deoxynucleotide building block of DNA.
  • Can be utilized as a substrate for UMP/CMP kinase.
  • Supports DNA biosynthesis.
  • Related to metabolic disease research.
Catalog No. 50-004-51665 Supplier Medchemexpress LLC Supplier No. HYW0092161G
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be utilized as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thereby supporting DNA biosynthesis.
  • Endogenous metabolite.
  • Deoxynucleotide building block of DNA.
  • Can be utilized as a substrate for UMP/CMP kinase.
  • Supports DNA biosynthesis.
  • Related to metabolic disease research.