Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate | 1032-65-1 | MFCD00006546 | 99.99% | 307.20 | 100 MG
SDP
Catalog No. 5000451427
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It serves as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis. This compound is intended for research use only and is not sold to patients.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis
Catalog No. 50-004-51427 Supplier Medchemexpress LLC Supplier No. HYW009216R100MG
2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It serves as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis. This compound is intended for research use only and is not sold to patients.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis