Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate-15N3 | 1032-65-1 | MFCD00006546 | 310.18 | 5 MG
SDP

Catalog No. 5000453225
Encompass_Preferred

2'-Deoxycytidine-5'-monophosphoric acid-15N3 is the 15N labeled form of 2'-Deoxycytidine-5'-monophosphoric acid, which is an endogenous metabolite.

  • Target: endogenous metabolite
  • Pathway: metabolic enzyme/protease
  • Storage: solution, -20°C, protect from light, stable for 2 years
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs

Catalog No. 50-004-53225 Supplier Medchemexpress LLC Supplier No. HYW009216S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxycytidine-5'-monophosphoric acid-15N3 is the 15N labeled form of 2'-Deoxycytidine-5'-monophosphoric acid, which is an endogenous metabolite.

  • Target: endogenous metabolite
  • Pathway: metabolic enzyme/protease
  • Storage: solution, -20°C, protect from light, stable for 2 years
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.