Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxycytidine-5'-monophosphate disodium salt, Thermo Scientific Chemicals

Catalog No. AAJ6572403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6572403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6572403 Supplier Thermo Scientific Chemicals Supplier No. J6572403
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13085-50-2 | Molecular Formula: C9H14N3Na2O7P | Molecular Weight: 353.178 g/mol

2?-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine 5-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

Solubility
Soluble in 200 mg plus 4 mL water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13085-50-2
Molecular Formula C9H14N3Na2O7P
Molecular Weight (g/mol) 353.178
MDL Number MFCD00069774
InChI Key ACIYKIIBZKDNAD-CDNBRZBRSA-N
Synonym 2'-deoxycytidine-5'-monophosphate disodium salt
PubChem CID 131846647
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O.[Na].[Na]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in 200 mg plus 4 mL water.
Formula Weight 351.16
Physical Form Powder
Chemical Name or Material 2'-Deoxycytidine-5'-monophosphate disodium salt
EINECSNumber 235-995-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.