Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphate (Standard) | 1032-65-1 | 99.6% | 307.20 | 10 MG
SDP
Catalog No. 5000451666
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphoric acid (Standard) is the analytical standard of 2'-Deoxycytidine-5'-monophosphoric acid. This product is intended for research and analytical applications. It is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis
  • Used in HPLC, GC, and MS research
Catalog No. 50-004-51666 Supplier Medchemexpress LLC Supplier No. HYW009216R10MG
2'-Deoxycytidine-5'-monophosphoric acid (Standard) is the analytical standard of 2'-Deoxycytidine-5'-monophosphoric acid. This product is intended for research and analytical applications. It is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Supports DNA biosynthesis
  • Used in HPLC, GC, and MS research