Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphoric acid | 1032-65-1 | 99.99% | 307.20 | 100 MG
SDP
Catalog No. 5000366835
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphate (also known as 2'-Deoxycytidine-5'-monophosphoric acid) is an endogenous metabolite. It is a deoxynucleotide building block that constitutes DNA. It can serve as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thus supporting DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis
Catalog No. 50-003-66835 Supplier Medchemexpress LLC Supplier No. HYW009216100MG
2'-Deoxycytidine-5'-monophosphate (also known as 2'-Deoxycytidine-5'-monophosphoric acid) is an endogenous metabolite. It is a deoxynucleotide building block that constitutes DNA. It can serve as a substrate for UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, thus supporting DNA biosynthesis.
  • Endogenous metabolite
  • Deoxynucleotide building block of DNA
  • Can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis