Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphoric acid-13C9,15N3 dilithium | 1032-65-1 | 99.8% | 330.98 | 10 MG
SDP

Catalog No. 5000366916
Encompass_Preferred

2'-Deoxycytidine-5'-monophosphoric acid-13C9,15N3 dilithium is a 13C and 15N-labeled version of 2'-Deoxycytidine-5'-monophosphoric acid, which is an endogenous metabolite.

  • 13C and 15N-labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Endogenous metabolite.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.

Catalog No. 50-003-66916 Supplier Medchemexpress LLC Supplier No. HYW009216S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxycytidine-5'-monophosphoric acid-13C9,15N3 dilithium is a 13C and 15N-labeled version of 2'-Deoxycytidine-5'-monophosphoric acid, which is an endogenous metabolite.

  • 13C and 15N-labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Endogenous metabolite.
  • Stable heavy isotopes of hydrogen, carbon, and other elements can be incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.