Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphoric acid-¹³C₉ (dilithium) | 99.5% | 328.00 | 1 MG
SDP

Catalog No. 5000452626
Encompass_Preferred

This product is 2'-Deoxycytidine-5'-monophosphate-¹³C₉ (dilithium), an isotopically labeled deoxycytidine monophosphate. It is provided as a colorless to light yellow solution at a concentration of 1 mg (70 mM in 43.55 μL Tris HCl/H₂O buffer pH=7.5). The compound is suitable for research applications requiring a high purity labeled standard.

  • Isotopically labeled with ¹³C₉
  • High purity of 99.5% (HPLC)
  • Isotopic enrichment of 98.9%
  • Provided in a solution format
  • Recommended for storage at -20°C for up to 2 years

Catalog No. 50-004-52626 Supplier Medchemexpress LLC Supplier No. HYW009216S41MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is 2'-Deoxycytidine-5'-monophosphate-¹³C₉ (dilithium), an isotopically labeled deoxycytidine monophosphate. It is provided as a colorless to light yellow solution at a concentration of 1 mg (70 mM in 43.55 μL Tris HCl/H₂O buffer pH=7.5). The compound is suitable for research applications requiring a high purity labeled standard.

  • Isotopically labeled with ¹³C₉
  • High purity of 99.5% (HPLC)
  • Isotopic enrichment of 98.9%
  • Provided in a solution format
  • Recommended for storage at -20°C for up to 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.