Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-monophosphoric acid (Standard) | 1032-65-1 | 99.6% | 307.20 | 50 MG
SDP
Catalog No. 5000366847
Encompass_Preferred
2'-Deoxycytidine-5'-monophosphoric acid (Standard) is the analytical standard of 2'-Deoxycytidine-5'-monophosphoric acid. This product is intended for research and analytical applications. 2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis.
  • Used as an analytical standard.
  • Supports DNA biosynthesis.
  • Suitable for qualitative research experiments.
  • Suitable for quantitative research experiments.
  • Suitable for methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-66847 Supplier Medchemexpress LLC Supplier No. HYW009216R50MG
2'-Deoxycytidine-5'-monophosphoric acid (Standard) is the analytical standard of 2'-Deoxycytidine-5'-monophosphoric acid. This product is intended for research and analytical applications. 2'-Deoxycytidine-5'-monophosphate is an endogenous metabolite and a deoxynucleotide building block of DNA. It can be used as a substrate of UMP/CMP kinase to form dCDP upon phosphorylation to dCTP, supporting DNA biosynthesis.
  • Used as an analytical standard.
  • Supports DNA biosynthesis.
  • Suitable for qualitative research experiments.
  • Suitable for quantitative research experiments.
  • Suitable for methodological research experiments in HPLC, GC, and MS.