Learn More
Medchemexpress LLC 2'-deoxycytidine-5'-triphosphate | 2056-98-6 | MFCD00135938 | 98.2% | 467.16 g/mol | C9H16N3O13P3 | 5 MG
SDP
Catalog No. 5000213064
Encompass_Preferred
Deoxycytidine triphosphate (dCTP) is a deoxynucleoside triphosphate used as a substrate for DNA polymerases in molecular biology workflows. It supplies the deoxycytidine nucleotide required for enzymatic DNA synthesis and is commonly used in PCR, quantitative PCR, cDNA synthesis, and DNA sequencing.
  • High chemical purity (≈98.2%)
  • Molecular weight 467.16 g/mol
  • Provides deoxycytidine nucleotide for polymerase-catalyzed DNA synthesis
  • Suitable for PCR, qPCR, cDNA synthesis, and DNA sequencing
  • Available in small lyophilized pack sizes (e.g., 5 mg) for laboratory workflows
  • Documented CAS number for unambiguous identification (2056-98-6)
Catalog No. 50-002-13064 Supplier Medchemexpress LLC Supplier No. HY1014005MG
Deoxycytidine triphosphate (dCTP) is a deoxynucleoside triphosphate used as a substrate for DNA polymerases in molecular biology workflows. It supplies the deoxycytidine nucleotide required for enzymatic DNA synthesis and is commonly used in PCR, quantitative PCR, cDNA synthesis, and DNA sequencing.
  • High chemical purity (≈98.2%)
  • Molecular weight 467.16 g/mol
  • Provides deoxycytidine nucleotide for polymerase-catalyzed DNA synthesis
  • Suitable for PCR, qPCR, cDNA synthesis, and DNA sequencing
  • Available in small lyophilized pack sizes (e.g., 5 mg) for laboratory workflows
  • Documented CAS number for unambiguous identification (2056-98-6)