Learn More
Medchemexpress LLC 2'-Deoxycytidine-5'-triphosphate-d14 dilithium | 2687960-70-7 | 95.0% | 493.11 | C9D14Li2N3O13P3 | 1 MG
SDP
Catalog No. 5000211145
Encompass_Preferred
Isotopically labeled deoxycytidine triphosphate-d14 supplied as the dilithium salt for use in DNA synthesis and analytical workflows. It serves as a deuterium-labeled nucleotide for enzymatic incorporation, tracer studies, and quantitative mass spectrometry.
  • Isotope-labeled for use as an internal standard and tracer in analytical workflows.
  • Suitable for enzymatic DNA synthesis, PCR, and cDNA synthesis.
  • Supplied as the dilithium salt to support aqueous solubility and handling.
  • High purity consistent with analytical and molecular biology applications.
  • Available in small-scale quantities for method development and validation.
Catalog No. 50-002-11145 Supplier Medchemexpress LLC Supplier No. HY101400S1MG
Isotopically labeled deoxycytidine triphosphate-d14 supplied as the dilithium salt for use in DNA synthesis and analytical workflows. It serves as a deuterium-labeled nucleotide for enzymatic incorporation, tracer studies, and quantitative mass spectrometry.
  • Isotope-labeled for use as an internal standard and tracer in analytical workflows.
  • Suitable for enzymatic DNA synthesis, PCR, and cDNA synthesis.
  • Supplied as the dilithium salt to support aqueous solubility and handling.
  • High purity consistent with analytical and molecular biology applications.
  • Available in small-scale quantities for method development and validation.