Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2′-deoxycytidine-5′-triphosphate dilithium salt | 2687960-70-7 | 95.7% | 493.11 g/mol | C9D14Li2N3O13P3 | 5 MG
SDP

Catalog No. 5000211146
Encompass_Preferred

Deuterium-labeled deoxycytidine triphosphate in dilithium form supplied for use as an isotopic internal standard and tracer in analytical and biochemical workflows. It is intended for applications requiring an isotopically labeled nucleotide reference such as mass spectrometry, quantitative assays, and tracer studies.

  • Deuterium-labeled nucleotide for isotopic tracing.
  • Provided as a dilithium salt, compatible with common solvents.
  • High purity suitable for analytical use (e.g., ~95.7%).
  • Molecular weight 493.11 g/mol; formula C9D14Li2N3O13P3.
  • Useful as an internal standard for mass spectrometry and quantitative assays.
  • Store according to supplier recommendations to maintain stability.

Catalog No. 50-002-11146 Supplier Medchemexpress LLC Supplier No. HY101400S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled deoxycytidine triphosphate in dilithium form supplied for use as an isotopic internal standard and tracer in analytical and biochemical workflows. It is intended for applications requiring an isotopically labeled nucleotide reference such as mass spectrometry, quantitative assays, and tracer studies.

  • Deuterium-labeled nucleotide for isotopic tracing.
  • Provided as a dilithium salt, compatible with common solvents.
  • High purity suitable for analytical use (e.g., ~95.7%).
  • Molecular weight 493.11 g/mol; formula C9D14Li2N3O13P3.
  • Useful as an internal standard for mass spectrometry and quantitative assays.
  • Store according to supplier recommendations to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.