Learn More
Medchemexpress LLC 2'-Deoxycytidine | 951-77-9 | 100.0% | 5 G
SDP
Catalog No. 5000410191
Encompass_Preferred
2'-Deoxycytidine, a deoxyribonucleoside, can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Deoxyribonucleoside
  • Inhibits biological effects of bromodeoxyuridine (Brdu)
  • Essential for the synthesis of nucleic acids
  • Can be used for cancer research
  • Inhibits tumor growth in mice
Catalog No. 50-004-10191 Supplier Medchemexpress LLC Supplier No. HYD01845G
2'-Deoxycytidine, a deoxyribonucleoside, can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Deoxyribonucleoside
  • Inhibits biological effects of bromodeoxyuridine (Brdu)
  • Essential for the synthesis of nucleic acids
  • Can be used for cancer research
  • Inhibits tumor growth in mice