Learn More
Medchemexpress LLC 2'-Deoxycytidine | 951-77-9 | 98.0% | 500 MG
SDP
Catalog No. 5000410143
Encompass_Preferred
2'-Deoxycytidine is a deoxyribonucleoside that can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Inhibits biological effects of bromodeoxyuridine (Brdu).
  • Essential for nucleic acid synthesis.
  • Used in cancer research.
  • Target: endogenous metabolite.
Catalog No. 50-004-10143 Supplier Medchemexpress LLC Supplier No. HYD0184500MG
2'-Deoxycytidine is a deoxyribonucleoside that can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Inhibits biological effects of bromodeoxyuridine (Brdu).
  • Essential for nucleic acid synthesis.
  • Used in cancer research.
  • Target: endogenous metabolite.