Learn More
Medchemexpress LLC 2'-Deoxycytidine | 951-77-9 | 99.95% | 1 ML
SDP
Catalog No. 5000410189
Encompass_Preferred
2'-Deoxycytidine is a deoxyribonucleoside that can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Inhibits biological effects of Bromodeoxyuridine (Brdu)
  • Essential for nucleic acid synthesis
  • Can be used for cancer research
  • Intended for research use only
Catalog No. 50-004-10189 Supplier Medchemexpress LLC Supplier No. HYD018410MM/1ML
2'-Deoxycytidine is a deoxyribonucleoside that can inhibit the biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for cancer research.
  • Inhibits biological effects of Bromodeoxyuridine (Brdu)
  • Essential for nucleic acid synthesis
  • Can be used for cancer research
  • Intended for research use only