Learn More
Medchemexpress LLC 2'-Deoxycytidine | 951-77-9 | 99.95% | 10 G
SDP
Catalog No. 5000409922
Encompass_Preferred
2'-Deoxycytidine is a deoxyribonucleoside that inhibits the biological effects of Bromodeoxyuridine (Brdu). It is essential for nucleic acid synthesis and can be used for cancer research.
  • Inhibits the biological effects of bromodeoxyuridine (Brdu)
  • Essential for nucleic acid synthesis
  • Can be used for cancer research
  • For research use only
Catalog No. 50-004-09922 Supplier Medchemexpress LLC Supplier No. HYD018410G
2'-Deoxycytidine is a deoxyribonucleoside that inhibits the biological effects of Bromodeoxyuridine (Brdu). It is essential for nucleic acid synthesis and can be used for cancer research.
  • Inhibits the biological effects of bromodeoxyuridine (Brdu)
  • Essential for nucleic acid synthesis
  • Can be used for cancer research
  • For research use only