Learn More
Medchemexpress LLC 2-Deoxycytidine | 951-77-9 | MFCD23103479 | 99.95% | 25 MG
SDP
Catalog No. 5000411276
Encompass_Preferred
2-Deoxycytidine, a deoxyribonucleoside, can inhibit biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for the research of cancer.
  • Essential for nucleic acid synthesis.
  • Used in cancer research.
  • Deoxyribonucleoside.
  • Can inhibit biological effects of Bromodeoxyuridine (Brdu).
Catalog No. 50-004-11276 Supplier Medchemexpress LLC Supplier No. 25MGHYD0184R
2-Deoxycytidine, a deoxyribonucleoside, can inhibit biological effects of Bromodeoxyuridine (Brdu). It is essential for the synthesis of nucleic acids and can be used for the research of cancer.
  • Essential for nucleic acid synthesis.
  • Used in cancer research.
  • Deoxyribonucleoside.
  • Can inhibit biological effects of Bromodeoxyuridine (Brdu).